Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX Labcorp Fentanyl Urine Visual Test, 2X Positive Control

Catalog No. 22026239
Change view
Click to view available options
Detectable Analytes:
Fentanyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-026-239 Fentanyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-026-239 Supplier MEDTOX Supplier No. 102520
Add to Cart
Add to Cart

2X positive control solution designed for use with the Labcorp Fentanyl Urine Visual Test.

  • Positive control formulated specifically for the Labcorp Fentanyl Urine Visual Test.
  • 2X concentration to ensure reliable verification during testing procedures.
  • Supplied in a 5 mL bottle for convenient handling and application.
  • Supports consistent quality control and accurate test performance.

Specifications

Concentration 2X Positive
Detectable Analytes Fentanyl
Sample Type Urine
Type Positive Control
Level High
Quantity 1 Bottle
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.