Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX Labcorp Fentanyl Urine Visual Test

Catalog No. 22025713
Change view
Click to view available options
Detectable Analytes:
Fentanyl (FEN)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
22-025-713 Fentanyl (FEN)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-025-713 Supplier MEDTOX Supplier No. 606000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lateral flow competitive immunoassay for the qualitative detection of norfentanyl in human urine.

  • Designed for fast and reliable detection of fentanyl exposure - ideal for patient care where rapid screening is critical.
  • This FDA cleared test
  • Provides over 96% accuracy in detecting norfentanyl, the primary fentanyl metabolite
  • Offers an extended detection window of up to 48 hours, even after fentanyl itself is no longer detected
  • Simplifies the process to allow rapid testing to be done without any lab equipment
  • Manufactured in the US, the Labcorp Fentanyl Urine Visual Test leverages our 30 years of expertise in developing and manufacturing rapid drug screening tests

Specifications

CE Marker No
Clia Complexity Moderate (Prescription Use Only)
Detectable Analytes Fentanyl (FEN)
Sample Type Urine
Disposable Single-use
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.