Learn More
Midwest Scientific LabDoctor 12 Mini Microcentrifuge
SDP
Catalog No. NC1622028
Encompass
Unlike traditional mini centrifuges, the new LabDoctor 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included, unique COMBI-Rotor is all that is required for running 12 microtubes and 4 PCR strips simultaneously.
Catalog No. NC1622028 Supplier Midwest Scientific Supplier No. MF12
Unlike traditional mini centrifuges, the new LabDoctor 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included, unique COMBI-Rotor is all that is required for running 12 microtubes and 4 PCR strips simultaneously.