Learn More
HCL Labels Corrosive Acid with GHS Pictogram Cabinet Sign - 3.5" x 11" Adhesive Vinyl, 1EA
SDP
Catalog No. 501633510
Encompass_Preferred
Tapes and Labels, HCL Labels, Inc., Use this label to mark a storage drawer, cabinet or secondary containment tray. This is to signify the presence of a corrosive acid hazard. Includes the updated GHS Pictogram.
Catalog No. 50-163-3510 Supplier HCL Labels Supplier No. SSLX134
Tapes and Labels, HCL Labels, Inc., Use this label to mark a storage drawer, cabinet or secondary containment tray. This is to signify the presence of a corrosive acid hazard. Includes the updated GHS Pictogram.