Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Labelmaster Biohazard Label, 4 x 4 in, Fluorescent Paper, 500/Roll

Catalog No. 501920711
Encompass

These biohazard labels are designed to clearly mark potential infectious waste with the universally recognized biohazard symbol. They meet OSHA's marking requirement in 29 CFR part 1910.1030 for bloodborne pathogens, ensuring compliance for containers of regulated waste, refrigerators, freezers, and other materials storing or shipping potentially infectious substances.

  • Fluorescent paper material with hot melt rubber adhesive
  • Dimensions: 4 x 4 in (101.6 x 101.6 mm)
  • Sold in rolls of 500 labels
  • Minimum application temperature of 10°F
  • Service temperature range from -50°F to 120°F
  • Total thickness of 7.1 mil

Catalog No. 50-192-0711 Supplier MSC Supplier No. 39525712
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These biohazard labels are designed to clearly mark potential infectious waste with the universally recognized biohazard symbol. They meet OSHA's marking requirement in 29 CFR part 1910.1030 for bloodborne pathogens, ensuring compliance for containers of regulated waste, refrigerators, freezers, and other materials storing or shipping potentially infectious substances.

  • Fluorescent paper material with hot melt rubber adhesive
  • Dimensions: 4 x 4 in (101.6 x 101.6 mm)
  • Sold in rolls of 500 labels
  • Minimum application temperature of 10°F
  • Service temperature range from -50°F to 120°F
  • Total thickness of 7.1 mil

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.