Learn More
HCL Labels Skull And Crossbones - Toxic Pictogram GHS Symbol - 4" x 4" Adhesive Vinyl (Pack of 25)
SDP
Catalog No. 501633475
Encompass_Preferred
Tapes and Labels, HCL Labels, Inc., Use this label to identify hazards according to OSHA GHS Hazard Communication Standards. Permanent adhesive and strong lamination allow for extended durability
Catalog No. 50-163-3475 Supplier HCL Labels Supplier No. SSL1095VN004425
Tapes and Labels, HCL Labels, Inc., Use this label to identify hazards according to OSHA GHS Hazard Communication Standards. Permanent adhesive and strong lamination allow for extended durability