Learn More
Electron Microscopy Sciences Label & Adhesive Remover 4.5 OZ
SDP
Catalog No. 50949189
Encompass_Preferred
The first label and adhesive remover to really work. To use, gently rub the label, wipe away the excess material from the surface, wait for 30 seconds then simply peel off the label, leaving a clean surface.
Catalog No. 50-949-189 Supplier Electron Microscopy Sciences Supplier No. 7182701
The first label and adhesive remover to really work. To use, gently rub the label, wipe away the excess material from the surface, wait for 30 seconds then simply peel off the label, leaving a clean surface.