Learn More
Electron Microscopy Sciences Label & Adhesive Remover 4.5 OZ
SDP
Supplier:  Electron Microscopy Sciences 7182706
Encompass_Preferred
The first label and adhesive remover to really work. To use, gently rub the label, wipe away the excess material from the surface, wait for 30 seconds then simply peel off the label, leaving a clean surface.
Catalog No. 50-949-557