Learn More
Andwin Scientific LABEL CRYOBABIES 1000/BOX
SDP
Catalog No. NC9814870
Encompass
Label; Cryo-Babies; 1.28 x 0.5 in.; Rectangular, Pre-cut, Peel-off labels, Specially designed to withstand cryogenic freezer temperatures; Chemically inert polyvinyl labels that adhere to all plastics and other materials
Catalog No. NC9814870 Supplier Andwin Scientific Supplier No. L20151
Label; Cryo-Babies; 1.28 x 0.5 in.; Rectangular, Pre-cut, Peel-off labels, Specially designed to withstand cryogenic freezer temperatures; Chemically inert polyvinyl labels that adhere to all plastics and other materials