Learn More
Andwin Scientific Laser Tough-Tags 0.94 x 0.50", Orange, 2,975/pk
SDP
Catalog No. NC1660861
Encompass_Preferred
Tough-Tags are chemically inert laboratory labels that strongly adhere to microtubes, test tubes, storage boxes and microscope slides. They resist organic solvents and caustic agents.
Catalog No. NC1660861 Supplier Andwin Scientific Supplier No. TTSW2240O
Tough-Tags are chemically inert laboratory labels that strongly adhere to microtubes, test tubes, storage boxes and microscope slides. They resist organic solvents and caustic agents.