Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Label Dispenser with Slide Cutter, Wall Mount
SDP

Catalog No. 22022555
Change view
Click to view available options
Holds:
Tape
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
22-022-555 Tape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-022-555 Supplier Poltex Supplier No. LABELDISPW
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Holds and dispenses a roll of labels. Slide-cutter allows for easy label cutting if needed. Wall mount (screws/anchors included).

  • Holds and dispenses a roll of labels
  • Slide-cutter allows for easy label cutting if needed
  • Durable PETG construction
  • Mounts to wall (screws/anchors included)

Specifications

Product Type Tape Dispenser (Single-roll)
Color Clear
Length (Metric) 368.30 mm
Width (Metric) 165.10 mm
Height (Metric) 279.40 mm
Holds Tape
Material PETG
Mounting Wall Mount
No. of Compartments 1
Product Style Top Loading
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.