Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Tape Logic Do Not Stack Labels, 3x4in, 500/Roll

Catalog No. 501919061
Encompass

These shipping labels are designed to alert handlers not to stack packages during transit. Featuring a clear 'Do Not Stack In Transit' message, they utilize a pressure-sensitive permanent adhesive for secure application on various package types.

  • Message: 'Do Not Stack In Transit'
  • Material: Semi-gloss coated paper
  • Color: Red, white, and black
  • Dimensions: 3 × 4 in
  • Quantity: 500 labels per roll
  • Application: Multi-use on boxes, envelopes, mailers, and bags
  • Adhesive: Pressure-sensitive permanent

Catalog No. 50-191-9061 Supplier MSC Supplier No. 39586656
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These shipping labels are designed to alert handlers not to stack packages during transit. Featuring a clear 'Do Not Stack In Transit' message, they utilize a pressure-sensitive permanent adhesive for secure application on various package types.

  • Message: 'Do Not Stack In Transit'
  • Material: Semi-gloss coated paper
  • Color: Red, white, and black
  • Dimensions: 3 × 4 in
  • Quantity: 500 labels per roll
  • Application: Multi-use on boxes, envelopes, mailers, and bags
  • Adhesive: Pressure-sensitive permanent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.