Learn More
Med Vet International Pre-Printed Label Comply Barcode Blue Label Sterilized Blue Sterilization Label 1.58 X 2.85 Cm, 1269B, 1125/Roll, Each
Catalog No. 502830046
Encompass_Preferred
Strong label adhesive keeps label in place during sterilization, eliminating the need for relabeling Durable, lightweight applicator is easy to use Strong label adhesive keeps label in place during sterilization, eliminating the need for relabeling Each label is 1.58 x 2.85 centimeters; there are 1,125 labels in a roll Blue
Catalog No. 50-283-0046 Supplier Med Vet International Supplier No. 270661EA
Strong label adhesive keeps label in place during sterilization, eliminating the need for relabeling Durable, lightweight applicator is easy to use Strong label adhesive keeps label in place during sterilization, eliminating the need for relabeling Each label is 1.58 x 2.85 centimeters; there are 1,125 labels in a roll Blue