Learn More
Med Vet International Pre-Printed Label Comply Laboratory Use Red Label Sterilized Red Sterilization Label 5/8 X 1-1/8 inch, 1269R, 1125/Roll, 1 Roll
Catalog No. 502830040
Encompass_Preferred
For use with the 3M Comply printing label applicator: Easily record critical information with these adhesive labels Strong label adhesive keeps label in place during sterilization, eliminating the need for relabeling Red font on white background Labels are 5/8 x 1-1/8 inches 1,125 labels per roll
Catalog No. 50-283-0040 Supplier Med Vet International Supplier No. 232435RL
For use with the 3M Comply printing label applicator: Easily record critical information with these adhesive labels Strong label adhesive keeps label in place during sterilization, eliminating the need for relabeling Red font on white background Labels are 5/8 x 1-1/8 inches 1,125 labels per roll