Learn More
Uline International Safe Handling Labels - Red Arrows, 3 x 4""; 500 labels per roll;
Catalog No. NC1765314
Encompass_Preferred
International Safe Handling Labels - Red Arrows, 3 x 4"; 500 labels per roll; Red arrows comply with IATA regulations. DIMENSIONS:Core: 3"; MATERIAL: Adhesive: PermanentTEMPERATURE: Application: 40 - 150 F; Performance: -30 to 150
Catalog No. NC1765314 Supplier Uline Supplier No. S850
International Safe Handling Labels - Red Arrows, 3 x 4"; 500 labels per roll; Red arrows comply with IATA regulations. DIMENSIONS:Core: 3"; MATERIAL: Adhesive: PermanentTEMPERATURE: Application: 40 - 150 F; Performance: -30 to 150