Learn More
Scinomix Inc LABEL KIT - 0.39"X1.2
SDP
Catalog No. NC1926268
Encompass
tube label + ribbons kit, 5,000 labels + ribbonWhite label with transparent patch on clear backing, no perforation2 rolls of Labels: 0.39" x 1.2" (10mm x 30mm), 1.55" gap, 3" core, 2,500L/r (2000-198)Ribbon: Premium Resin Ribbon, 40mm x 450m, 1" ID (2000-464)
Catalog No. NC1926268 Supplier Scinomix Inc Supplier No. 20001980
tube label + ribbons kit, 5,000 labels + ribbonWhite label with transparent patch on clear backing, no perforation2 rolls of Labels: 0.39" x 1.2" (10mm x 30mm), 1.55" gap, 3" core, 2,500L/r (2000-198)Ribbon: Premium Resin Ribbon, 40mm x 450m, 1" ID (2000-464)