Learn More
HCL Labels Danger - Peroxide Forming Chemical - 2" x 3" Adhesive Vinyl (Pack of 100)
SDP
Catalog No. 501633515
Encompass_Preferred
Tapes and Labels, HCL Labels, Inc., For peroxide forming chemicals. Label allows to record arrival, opened, expiration and disposal dates.
Catalog No. 50-163-3515 Supplier HCL Labels Supplier No. SHL0059VN0023100
Tapes and Labels, HCL Labels, Inc., For peroxide forming chemicals. Label allows to record arrival, opened, expiration and disposal dates.