Learn More
HCL Labels Danger - Peroxide Forming Chemical - 3" x 5" Adhesive Vinyl (Pack of 25)
SDP
Catalog No. 501633518
Encompass_Preferred
Tapes and Labels, HCL Labels, Inc., For peroxide forming chemicals. Label allows to record arrival, opened, expiration and disposal dates.
Catalog No. 50-163-3518 Supplier HCL Labels Supplier No. SHL0059VN003525
Tapes and Labels, HCL Labels, Inc., For peroxide forming chemicals. Label allows to record arrival, opened, expiration and disposal dates.