Learn More
HCL Labels Biohazard - Liquid Biowaste Adhesive Vinyl Label - 2" x 3" (Pack of 100)
SDP
Catalog No. 501633420
Encompass_Preferred
Tapes and Labels, HCL Labels, Inc., This is an ideal label to identify beakers and containers holding liquid biowaste generated during laboratory operations. Permanent adhesive and strong lamination allow for extended durability.
Catalog No. 50-163-3420 Supplier HCL Labels Supplier No. SSL1704VN0023100
Tapes and Labels, HCL Labels, Inc., This is an ideal label to identify beakers and containers holding liquid biowaste generated during laboratory operations. Permanent adhesive and strong lamination allow for extended durability.