Learn More
Brady™ Label Tray for BradyJet J7300 Printer, Gray
Catalog No. 18038234 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
BradyJet J7300 Printer
Catalog No. For Use With (Equipment)
18-038-234 BradyJet J7300 Printer
1 options
Catalog No. 18-038-234 Supplier Brady™ Supplier No. JTRAY1
Add to Cart
Add to Cart
Durable label tray ensures smooth operation by catching printed labels after cutting.
  • Extends out for easy use and retracts flush with the printer for secure storage.
  • Periodically remove labels from tray after cutting to avoid a build-up, which can jam the cutter.
  • The dimensions are (HxWxD) 0.75 x 6 x 18.5 in.
Specifications
Product Type Replacement Catch Tray
For Use With (Equipment) BradyJet J7300 Printer