Learn More
Electron Microscopy Sciences Label with HMIS warning, 0.875 x 0.875"
SDP
Catalog No. 501930448
Encompass_Preferred
These warning labels will withstand multiple freeze-thaw cycles, boiling water and autoclaving, dry ice, UV rays, Gamma irrradiation, most chemicals and solvents, and other harsh laboratory environments. They are ideal for prolonged storage inside liquid nitrogen tanks, freezers and dry ice as well as for transportation of hazardous biological samples and other hazardous materials in dry ice containers, portable freezers and portable liquid nitrogen tanks. The pictograms on the laminated labels are protected against water, alcohols, chemicals, and solvents. Temperature range from -196 deg C to +100 deg C.
Size: 0.875 x 0.875"
Perforated: Yes
Laminated: Yes
Catalog No. 50-193-0448 Supplier Electron Microscopy Sciences Supplier No. 7742912
These warning labels will withstand multiple freeze-thaw cycles, boiling water and autoclaving, dry ice, UV rays, Gamma irrradiation, most chemicals and solvents, and other harsh laboratory environments. They are ideal for prolonged storage inside liquid nitrogen tanks, freezers and dry ice as well as for transportation of hazardous biological samples and other hazardous materials in dry ice containers, portable freezers and portable liquid nitrogen tanks. The pictograms on the laminated labels are protected against water, alcohols, chemicals, and solvents. Temperature range from -196 deg C to +100 deg C.
Size: 0.875 x 0.875"
Perforated: Yes
Laminated: Yes