Learn More
Grainger Brady Pre-Cut Label Stock Roll, Polyester, Rectangle, White, 5,000/Roll
Catalog No. 502777888
Encompass_Preferred
These pre-cut rectangle labels are designed for reliable laboratory identification of samples stored in extreme conditions. Constructed from durable, thermal transfer-printable white polyester, these labels feature an aggressive permanent acrylic adhesive capable of being applied to frozen surfaces, including glass and polypropylene. They are suitable for use in liquid nitrogen and freezer environments, providing excellent resistance to chemicals and smearing.
  • Designed for laboratory applications including vials, centrifuge tubes, test tubes, straws, and slides
  • Service temperature range of -320°F to 230°F
  • Can be applied to frozen surfaces
  • Chemical and smear-resistant for long-term durability
  • Compatible with thermal transfer printers
  • Compliant with 2011/65/EU and 2015/863 RoHS standards
Catalog No. 50-277-7888 Supplier Grainger Supplier No. 18DU49
These pre-cut rectangle labels are designed for reliable laboratory identification of samples stored in extreme conditions. Constructed from durable, thermal transfer-printable white polyester, these labels feature an aggressive permanent acrylic adhesive capable of being applied to frozen surfaces, including glass and polypropylene. They are suitable for use in liquid nitrogen and freezer environments, providing excellent resistance to chemicals and smearing.
  • Designed for laboratory applications including vials, centrifuge tubes, test tubes, straws, and slides
  • Service temperature range of -320°F to 230°F
  • Can be applied to frozen surfaces
  • Chemical and smear-resistant for long-term durability
  • Compatible with thermal transfer printers
  • Compliant with 2011/65/EU and 2015/863 RoHS standards