Learn More
Grainger Brady FreezerBondz Ultra-Thin Cryogenic Polyester Laboratory Labels, 0.5 in. Circle, 3,000/Roll
Catalog No. 502777510
Encompass_Preferred
These ultra-thin, cryogenic polyester labels are engineered for laboratory identification on curved, frozen surfaces. Designed to maintain integrity in extreme environments, the labels can be applied directly to pre-frozen glass, plastic, and stainless steel containers. They feature an aggressive, permanent acrylic adhesive that ensures reliable bonding to vials, centrifuge samples, and tubes, even when wrapped with an overlap.
  • Withstands temperatures from -196°C to +110°C (-320°F to +230°F)
  • Chemical and smear resistant for long-term sample durability
  • Compatible with liquid nitrogen storage
  • Die-cut design with 0.5 in. diameter
  • Requires thermal transfer printing with compatible ribbons
Catalog No. 50-277-7510 Supplier Grainger Supplier No. 18CE70
These ultra-thin, cryogenic polyester labels are engineered for laboratory identification on curved, frozen surfaces. Designed to maintain integrity in extreme environments, the labels can be applied directly to pre-frozen glass, plastic, and stainless steel containers. They feature an aggressive, permanent acrylic adhesive that ensures reliable bonding to vials, centrifuge samples, and tubes, even when wrapped with an overlap.
  • Withstands temperatures from -196°C to +110°C (-320°F to +230°F)
  • Chemical and smear resistant for long-term sample durability
  • Compatible with liquid nitrogen storage
  • Die-cut design with 0.5 in. diameter
  • Requires thermal transfer printing with compatible ribbons