Learn More
Grainger Brady Thermal Transfer Labels, Polyester, FreezerBondz, 1 in. x 1 in., 3000/Roll
Catalog No. 502777513
Encompass_Preferred
These FreezerBondz™ polyester labels are engineered for demanding laboratory environments, providing reliable identification for samples stored in extreme cold, including liquid nitrogen. Constructed with an aggressive permanent acrylic adhesive, these white, die-cut labels can be applied directly to pre-frozen glass, plastic, and stainless steel surfaces. The durable material is designed to withstand harsh laboratory conditions, including chemical exposure and abrasions.
  • Temperature performance from -196°C to 100°C (-320°F to 212°F)
  • Ideal for curved surfaces such as vials, centrifuge tubes, and slides
  • Compatible with thermal transfer printing technology
  • Chemical and smear resistant for long-term durability
  • Supplied on 3 in. core rolls for use with industrial label printers
Catalog No. 50-277-7513 Supplier Grainger Supplier No. 18CY21
These FreezerBondz™ polyester labels are engineered for demanding laboratory environments, providing reliable identification for samples stored in extreme cold, including liquid nitrogen. Constructed with an aggressive permanent acrylic adhesive, these white, die-cut labels can be applied directly to pre-frozen glass, plastic, and stainless steel surfaces. The durable material is designed to withstand harsh laboratory conditions, including chemical exposure and abrasions.
  • Temperature performance from -196°C to 100°C (-320°F to 212°F)
  • Ideal for curved surfaces such as vials, centrifuge tubes, and slides
  • Compatible with thermal transfer printing technology
  • Chemical and smear resistant for long-term durability
  • Supplied on 3 in. core rolls for use with industrial label printers