Learn More
Grainger Brady StainerBondz Laboratory Labels, Satin Polyester, 0.2 in x 2.5 in, 2500/Roll
Catalog No. 502777775
Encompass_Preferred
These StainerBondz™ microscope slide labels are engineered to deliver high performance in demanding laboratory environments. Designed for use with thermal transfer printers, these die-cut polyester labels remain legible and securely attached throughout rigorous slide staining processes, including exposure to Xylene, Ethanol, and Toluene. The labels feature a permanent acrylic adhesive suitable for bonding to smooth or textured glass surfaces.
  • Engineered for harsh environments and H&E staining processes
  • Resists a wide range of chemicals, oils, and water
  • Maintains performance across temperatures from -112°F to 266°F (-80°C to 130°C)
  • Features permanent acrylic adhesive for secure bonding to glass
  • Compatible with 3 in. core thermal transfer printers
  • RoHS compliant to 2011/65/EU and 2015/863
  • Includes 2,500 labels per roll
Catalog No. 50-277-7775 Supplier Grainger Supplier No. 18CE68
These StainerBondz™ microscope slide labels are engineered to deliver high performance in demanding laboratory environments. Designed for use with thermal transfer printers, these die-cut polyester labels remain legible and securely attached throughout rigorous slide staining processes, including exposure to Xylene, Ethanol, and Toluene. The labels feature a permanent acrylic adhesive suitable for bonding to smooth or textured glass surfaces.
  • Engineered for harsh environments and H&E staining processes
  • Resists a wide range of chemicals, oils, and water
  • Maintains performance across temperatures from -112°F to 266°F (-80°C to 130°C)
  • Features permanent acrylic adhesive for secure bonding to glass
  • Compatible with 3 in. core thermal transfer printers
  • RoHS compliant to 2011/65/EU and 2015/863
  • Includes 2,500 labels per roll