Learn More
Grainger BRADY Pre-Cut Label Stock Roll, Rectangle, White Polyester, 3,000/Roll
Catalog No. 502777724
Encompass_Preferred
These cryogenic, thermal transfer printable polyester labels are engineered for laboratory identification applications. Featuring an aggressive permanent acrylic adhesive, they are suitable for labeling vials, centrifuge tubes, test tubes, straws, and slides, with the ability to be applied directly to already-frozen surfaces.
  • Designed for use in temperatures ranging from -196°C (-320°F) to +130°C (+266°F)
  • Compatible with liquid nitrogen and freezer storage, including cycles between deep freeze and boiling water
  • Constructed from durable matte white polyester with high-performance adhesive
  • Compatible with thermal transfer print technology
  • RoHS compliant and free of substances of very high concern (SVHC)
Catalog No. 50-277-7724 Supplier Grainger Supplier No. 18CT61
These cryogenic, thermal transfer printable polyester labels are engineered for laboratory identification applications. Featuring an aggressive permanent acrylic adhesive, they are suitable for labeling vials, centrifuge tubes, test tubes, straws, and slides, with the ability to be applied directly to already-frozen surfaces.
  • Designed for use in temperatures ranging from -196°C (-320°F) to +130°C (+266°F)
  • Compatible with liquid nitrogen and freezer storage, including cycles between deep freeze and boiling water
  • Constructed from durable matte white polyester with high-performance adhesive
  • Compatible with thermal transfer print technology
  • RoHS compliant and free of substances of very high concern (SVHC)