Learn More
Grainger Brady Laboratory Labels, FreezerBondz Cryogenic Polyester, 0.5 in x 1 in, 3,000/Roll
Catalog No. 502778046
Encompass_Preferred
These ultra-thin cryogenic polyester laboratory labels are engineered for exceptional durability in challenging cryogenic environments. Designed with an aggressive, permanent acrylic adhesive, they allow for direct application to pre-frozen glass, plastic, and stainless steel surfaces. Each die-cut label features a white finish and includes a vial top for comprehensive identification. These labels are resistant to chemicals and abrasions, making them ideal for vials, tubes, slides, and other laboratory samples stored in liquid nitrogen or extreme temperature conditions.
  • Withstands temperatures from -320°F to 230°F (-196°C to 110°C)
  • Adheres to curved and pre-frozen surfaces
  • Compatible with thermal transfer printers using a 3 in. core
  • Resistant to chemicals and smearing
  • Features a permanent acrylic adhesive for secure bonding
  • Includes a circular vial top label
Catalog No. 50-277-8046 Supplier Grainger Supplier No. 18DC15
These ultra-thin cryogenic polyester laboratory labels are engineered for exceptional durability in challenging cryogenic environments. Designed with an aggressive, permanent acrylic adhesive, they allow for direct application to pre-frozen glass, plastic, and stainless steel surfaces. Each die-cut label features a white finish and includes a vial top for comprehensive identification. These labels are resistant to chemicals and abrasions, making them ideal for vials, tubes, slides, and other laboratory samples stored in liquid nitrogen or extreme temperature conditions.
  • Withstands temperatures from -320°F to 230°F (-196°C to 110°C)
  • Adheres to curved and pre-frozen surfaces
  • Compatible with thermal transfer printers using a 3 in. core
  • Resistant to chemicals and smearing
  • Features a permanent acrylic adhesive for secure bonding
  • Includes a circular vial top label