Learn More
Grainger Brady StainerBondz Laboratory Labels, Polyester, 0.75 in x 0.9 in, 3,000/Roll
Catalog No. 502778543
Encompass_Preferred
These StainerBondz™ laboratory labels are engineered from high-performance, satin-finish polyester specifically for microscope slide identification. The labels are designed to remain legible and securely attached throughout demanding slide staining processes, including exposure to xylene, ethanol, and toluene. Featuring a permanent acrylic adhesive, they provide a reliable bond to glass surfaces even under extreme temperature conditions.
  • Withstand harsh laboratory environments and chemical exposure including H&E staining
  • Temperature performance range of -80°C to +130°C (-112°F to 266°F)
  • Compatible with thermal transfer printing technology
  • Die-cut design with rounded corners for precision
  • Resistant to chemicals, oils, and water
Catalog No. 50-277-8543 Supplier Grainger Supplier No. 18DC52
These StainerBondz™ laboratory labels are engineered from high-performance, satin-finish polyester specifically for microscope slide identification. The labels are designed to remain legible and securely attached throughout demanding slide staining processes, including exposure to xylene, ethanol, and toluene. Featuring a permanent acrylic adhesive, they provide a reliable bond to glass surfaces even under extreme temperature conditions.
  • Withstand harsh laboratory environments and chemical exposure including H&E staining
  • Temperature performance range of -80°C to +130°C (-112°F to 266°F)
  • Compatible with thermal transfer printing technology
  • Die-cut design with rounded corners for precision
  • Resistant to chemicals, oils, and water