Learn More
Grainger Brady Self-Laminating Cryogenic Laboratory Labels, Polyester, 1 in. x 5.25 in., 2,000/Pk.
Catalog No. 502776140
Encompass_Preferred
These self-laminating cryogenic polyester labels are designed to provide durable identification for laboratory samples stored in extreme conditions. Featuring a clear overlaminate, the labels protect printed text, barcodes, and graphics from solvents, smudges, and temperature extremes, making them suitable for use with vials, centrifuge tubes, test tubes, straws, and slides. The labels utilize a permanent acrylic adhesive for reliable bonding to smooth surfaces including glass, stainless steel, and copper.
  • Temperature range: -321°F to 230°F (-196°C to 110°C)
  • Durable, chemical-resistant matte polyester construction
  • Self-laminating design with clear overlaminate for print protection
  • Compatible with 3 in. core thermal transfer printers
  • Permanent acrylic adhesive ensures secure attachment to smooth surfaces
  • Suitable for cryogenic and autoclave applications
Catalog No. 50-277-6140 Supplier Grainger Supplier No. 18CE11
These self-laminating cryogenic polyester labels are designed to provide durable identification for laboratory samples stored in extreme conditions. Featuring a clear overlaminate, the labels protect printed text, barcodes, and graphics from solvents, smudges, and temperature extremes, making them suitable for use with vials, centrifuge tubes, test tubes, straws, and slides. The labels utilize a permanent acrylic adhesive for reliable bonding to smooth surfaces including glass, stainless steel, and copper.
  • Temperature range: -321°F to 230°F (-196°C to 110°C)
  • Durable, chemical-resistant matte polyester construction
  • Self-laminating design with clear overlaminate for print protection
  • Compatible with 3 in. core thermal transfer printers
  • Permanent acrylic adhesive ensures secure attachment to smooth surfaces
  • Suitable for cryogenic and autoclave applications