Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Decker Tape Explosive Label, 4 x 4 in, 500 ea

Catalog No. 501918826
Encompass

This D.O.T. CLASS 1 EXPLOSIVE 1.3C label is designed for clear hazard identification, featuring a permanent adhesive and supplied on rolls for efficient application in industrial and commercial settings.

  • Size: 4 x 4 in
  • Labels per roll: 500
  • Material: Bright orange paper
  • Ink color: Black
  • Adhesive: Permanent
  • Core size: 1.5 in ID
  • Manufactured in the USA

Catalog No. 50-191-8826 Supplier MSC Supplier No. 39584206
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This D.O.T. CLASS 1 EXPLOSIVE 1.3C label is designed for clear hazard identification, featuring a permanent adhesive and supplied on rolls for efficient application in industrial and commercial settings.

  • Size: 4 x 4 in
  • Labels per roll: 500
  • Material: Bright orange paper
  • Ink color: Black
  • Adhesive: Permanent
  • Core size: 1.5 in ID
  • Manufactured in the USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.