Learn More
Essendant LABEL7/8X2/8REM825WH
Supplier:  Essendant AVE61501
Encompass_Preferred
Labels feature a special adhesive that s engineered to stick strong and remove cleanly from smooth hard surfaces including metal plastic and glass The durable professional-grade material resists water chemicals and abrasion Also provides superior barcode scanning Great for use in warehouses offices factories hospitality and more The removable adhesive sticks securely but removes cleanly for easy on-demand changes Simple to customize with text graphics or barcodes using the free software from Avery Design and Print Recommended for indoor use Not recommended for use on walls Compatible with laser and inkjet printers Shape- Rectangular Label Size - text- 0 88x2 63 Label Color(s)- White Color Family- White
Catalog No. 50-000-21982