Learn More
Research Products International Corp LABeler, Standard Label Tape Cassette, 12mm, 26 Feet, Clear with Black Print
SDP
Catalog No. 501367669
Encompass
Standard LABeler tape withstands autoclaving, boiling, freezing applications as low as -50°C.
Catalog No. 50-136-7669 Supplier Research Products International Corp Supplier No. 165104
Standard LABeler tape withstands autoclaving, boiling, freezing applications as low as -50°C.