Learn More
Research Products International Corp LABeler, Standard Label Tape Cassette, 12mm, 26 Feet, Yellow with Black Print
SDP
Catalog No. 501367670
Encompass
Standard LABeler tape withstands autoclaving, boiling, freezing applications as low as -50°C.
Catalog No. 50-136-7670 Supplier Research Products International Corp Supplier No. 165105
Standard LABeler tape withstands autoclaving, boiling, freezing applications as low as -50°C.