Learn More
Research Products International Corp LABeler, Standard Label Tape Cassette, 6mm, 26 Feet, White with Black Print
SDP
Catalog No. 501367666
Encompass
Standard LABeler tape withstands autoclaving, boiling, freezing applications as low as -50°C.
Catalog No. 50-136-7666 Supplier Research Products International Corp Supplier No. 165101
Standard LABeler tape withstands autoclaving, boiling, freezing applications as low as -50°C.