Learn More
Research Products International Corp LABeler, ULT Ultra-Low Temperature Label Tape Cassette, 26 Feet, 12mm, White with Black Print
SDP
Catalog No. 501367673
Encompass
Standard LABeler tape withstands autoclaving, boiling, freezing applications as low as -80°C.
Catalog No. 50-136-7673 Supplier Research Products International Corp Supplier No. 165108
Standard LABeler tape withstands autoclaving, boiling, freezing applications as low as -80°C.