Learn More
Agilent Technologies SIGNAL™ DMB LABELING KIT
Catalog No. NC9416968
Encompass
Convenient labeling of sialic acid with 1,2-diamino-4,5-methylenedioxybenzene (DMB) with subsequent identification by comparison with a Sialic Acid. Reference Panel. High sensitivity and specificity. Good resolution of O-acetylated derivatives. Minimal artifacts due to O-acetyl ester migration. from the 7- to the 9- position. Samples may contain up to 2.5 nmols sialic acid. Up to 20 individual samples (2 batches of 10).
Catalog No. NC9416968 Supplier Agilent Technologies Supplier No. GKK407
Convenient labeling of sialic acid with 1,2-diamino-4,5-methylenedioxybenzene (DMB) with subsequent identification by comparison with a Sialic Acid. Reference Panel. High sensitivity and specificity. Good resolution of O-acetylated derivatives. Minimal artifacts due to O-acetyl ester migration. from the 7- to the 9- position. Samples may contain up to 2.5 nmols sialic acid. Up to 20 individual samples (2 batches of 10).