Learn More
Fluidigm Corp Maxpar X8 Antibody Labeling Kit, 152Sm - 4 Rxn
SDP
Catalog No. NC1361657
Encompass
This Maxpar kit lets you conjugate IgG antibodies with the 152Sm metal isotope, with each kit containing enough material to conjugate 4-40 antibodies in 100 µg amounts.
Catalog No. NC1361657 Supplier Fluidigm Corp Supplier No. 201152A
This Maxpar kit lets you conjugate IgG antibodies with the 152Sm metal isotope, with each kit containing enough material to conjugate 4-40 antibodies in 100 µg amounts.