Learn More
Electron Microscopy Sciences Labeling Tape, 0.75" x 500", Blue 500 IN
SDP
Catalog No. 5034216
Encompass_Preferred
Labeling tape that is markable with a pen, pencil, or marker. Leaves no adhesive residue when peeled off. Water, oil, and acid resistant. Withstands cold (-23 degree C) and hot (121 degree C) temperatures.
Size: ¾" x 500"
Color: Blue
Catalog No. 50-342-16 Supplier Electron Microscopy Sciences Supplier No. 77627B
Labeling tape that is markable with a pen, pencil, or marker. Leaves no adhesive residue when peeled off. Water, oil, and acid resistant. Withstands cold (-23 degree C) and hot (121 degree C) temperatures.
Size: ¾" x 500"
Color: Blue