Learn More
Electron Microscopy Sciences Labeling Tape, 0.75"x 2160 ", Green 2160 IN
SDP
Catalog No. 5034218
Encompass_Preferred
Labeling tape that is markable with a pen, pencil, or marker. Leaves no adhesive residue when peeled off. Water, oil, and acid resistant. Withstands cold (-23 degree C) and hot (121 degree C) temperatures.
Size: ¾" x 2160 "
Color: Green
Catalog No. 50-342-18 Supplier Electron Microscopy Sciences Supplier No. 77628G
Labeling tape that is markable with a pen, pencil, or marker. Leaves no adhesive residue when peeled off. Water, oil, and acid resistant. Withstands cold (-23 degree C) and hot (121 degree C) temperatures.
Size: ¾" x 2160 "
Color: Green