Learn More
MSC Labelmaster Oxidizer Placard, Removable Vinyl, 273 mm, 25/Pk.
Catalog No. 5000677184
Encompass
These Oxidizer Worded Placards are designed for transporting oxidizing solids. These DOT placards comply with the requirements of 49 CFR 172.500 for both domestic and international hazardous material shipments via highway, rail, and water. They are made with UV inks that resist peeling, chipping, and fading. The placards are printed on Removable Vinyl, suitable for both long-term and short-term shipping, and can be removed with little to no residue. They are yellow and black, measure 273 x 273 mm, and are sold in packs of 25.
  • Complies with 49 CFR 172.500 for hazardous materials transport
  • UV inks resist peeling, chipping, and fading
  • Removable vinyl for long-term or short-term shipping
  • Removes with minimal residue
  • Designed for Hazard Class 5 oxidizing solids
  • Dimensions are 273 x 273 mm.
Catalog No. 50-006-77184 Supplier MSC Supplier No. 23118300
These Oxidizer Worded Placards are designed for transporting oxidizing solids. These DOT placards comply with the requirements of 49 CFR 172.500 for both domestic and international hazardous material shipments via highway, rail, and water. They are made with UV inks that resist peeling, chipping, and fading. The placards are printed on Removable Vinyl, suitable for both long-term and short-term shipping, and can be removed with little to no residue. They are yellow and black, measure 273 x 273 mm, and are sold in packs of 25.
  • Complies with 49 CFR 172.500 for hazardous materials transport
  • UV inks resist peeling, chipping, and fading
  • Removable vinyl for long-term or short-term shipping
  • Removes with minimal residue
  • Designed for Hazard Class 5 oxidizing solids
  • Dimensions are 273 x 273 mm.