Learn More
MSC Labelmaster Placard, 10-3/4 in. Square, Vinyl, Corrosive, 1 ea
Catalog No. 5000767674
Encompass
This vinyl placard is designed for displaying hazardous material information, measuring 10-3/4 in. wide by 10-3/4 in. high. It features a Corrosive legend and is suitable for various industrial applications requiring clear hazard identification.
  • Constructed from durable vinyl material
  • Features a black Corrosive legend for clear identification
  • Protected with a UV coating for enhanced durability
  • Complies with DOT 49 CFR 172.519 specifications
Catalog No. 50-007-67674 Supplier MSC Supplier No. 23117799
This vinyl placard is designed for displaying hazardous material information, measuring 10-3/4 in. wide by 10-3/4 in. high. It features a Corrosive legend and is suitable for various industrial applications requiring clear hazard identification.
  • Constructed from durable vinyl material
  • Features a black Corrosive legend for clear identification
  • Protected with a UV coating for enhanced durability
  • Complies with DOT 49 CFR 172.519 specifications