Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC LABELS AVE 5267 2000 PER PK

Catalog No. NC1365281
Encompass

These label maker labels are designed for use with laser printers, offering a practical solution for shipping and general labeling applications.

  • White paper material
  • Contains 2,000 labels per roll
  • For use with laser printers
  • Features 1/2 in. width
  • Features 1-3/4 in. length

Catalog No. NC1365281 Supplier MSC Supplier No. 81629313
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These label maker labels are designed for use with laser printers, offering a practical solution for shipping and general labeling applications.

  • White paper material
  • Contains 2,000 labels per roll
  • For use with laser printers
  • Features 1/2 in. width
  • Features 1-3/4 in. length

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.