Learn More
GA International NitroTAG – Cryogenic Barcode Labels for Liquid Nitrogen Storage
Catalog No. NC1571452
Encompass_Preferred
These labels are ideally suited for storage of small diameter cryo tubes and vials, polypropylene cryo/freezer boxes, cardboard freezer boxes, cell culture plates, and plastic bags in liquid and vapor phase liquid nitrogen (-196°C/-321°F), and lab freezers (-80°C/-112°F), as well as for dry ice transportation. Waterproof and made of a strong thermoplastic film, these labels can be immersed in liquid nitrogen immediately following application, and withstand multiple freeze-thaw cycles. Compatible with all common thermal-transfer printers, they can be printed on with alphanumeric text, barcodes, serial numbers, serial barcodes, and other types of information.
Catalog No. NC1571452 Supplier GA International Supplier No. JTTA62C31YE
These labels are ideally suited for storage of small diameter cryo tubes and vials, polypropylene cryo/freezer boxes, cardboard freezer boxes, cell culture plates, and plastic bags in liquid and vapor phase liquid nitrogen (-196°C/-321°F), and lab freezers (-80°C/-112°F), as well as for dry ice transportation. Waterproof and made of a strong thermoplastic film, these labels can be immersed in liquid nitrogen immediately following application, and withstand multiple freeze-thaw cycles. Compatible with all common thermal-transfer printers, they can be printed on with alphanumeric text, barcodes, serial numbers, serial barcodes, and other types of information.