Learn More
USA Scientific Inc Tough-Tags, 0.94 inch x 0.5 inch, red, on rolls
Catalog No. 507541506
Encompass
Tough-Tags, small, red labels for 0.5 mL tubes, 0.94 inch L x 0.5 inch W, 1000/roll. For -40C to +121C.
Catalog No. 50-754-1506 Supplier USA Scientific Inc Supplier No. 91881004
Tough-Tags, small, red labels for 0.5 mL tubes, 0.94 inch L x 0.5 inch W, 1000/roll. For -40C to +121C.