Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Labetalol | 36894-69-6 | 99.9% | 1 ML
SDP

Catalog No. 5000232931
Encompass_Preferred

Labetalol 10 mM in DMSO (1 mL) is a ready-to-use solution form of an α1/β-adrenergic receptor antagonist for in vitro pharmacology and receptor binding studies. It is provided with analytical documentation for research use.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied in a 1 mL vial suitable for small-scale assays.
  • High purity for biochemical and pharmacological applications.
  • Molecular weight 328.41 g/mol and formula C19H24N2O3.
  • Includes certificate of analysis and safety data sheet.
  • Intended for in vitro receptor binding and pharmacology studies.

Catalog No. 50-002-32931 Supplier Medchemexpress LLC Supplier No. HY12138310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labetalol 10 mM in DMSO (1 mL) is a ready-to-use solution form of an α1/β-adrenergic receptor antagonist for in vitro pharmacology and receptor binding studies. It is provided with analytical documentation for research use.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied in a 1 mL vial suitable for small-scale assays.
  • High purity for biochemical and pharmacological applications.
  • Molecular weight 328.41 g/mol and formula C19H24N2O3.
  • Includes certificate of analysis and safety data sheet.
  • Intended for in vitro receptor binding and pharmacology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.