Learn More
Apexbio Technology LLC Labetalol HCl 32780-64-6 200mg
SDP
Catalog No. 5000063927
Encompass_Preferred
Labetalol HCl (CAS 32780-64-6) is a small molecule antagonist targeting - and -adrenergic receptors It selectively inhibits -adrenergic receptors on vascular smooth muscle preventing adrenergic-mediated vasoconstriction Concurrently Labetalol HCl exhibits non-selective inhibition of -adrenergic receptors in cardiac and bronchial tissues reducing heart rate cardiac contractility cardiac output and arterial blood pressure Due to these vasoactive and cardioregulatory effects it is frequently utilized in research related to cardiovascular physiology hypertension and adrenergic signaling pathways
Catalog No. 50-000-63927 Supplier Apexbio Technology LLC Supplier No. B4795200
Labetalol HCl (CAS 32780-64-6) is a small molecule antagonist targeting - and -adrenergic receptors It selectively inhibits -adrenergic receptors on vascular smooth muscle preventing adrenergic-mediated vasoconstriction Concurrently Labetalol HCl exhibits non-selective inhibition of -adrenergic receptors in cardiac and bronchial tissues reducing heart rate cardiac contractility cardiac output and arterial blood pressure Due to these vasoactive and cardioregulatory effects it is frequently utilized in research related to cardiovascular physiology hypertension and adrenergic signaling pathways