Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Labetalol hydrochloride, 99%

Catalog No. AC462600250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462600250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462600250 Supplier Thermo Scientific Chemicals Supplier No. 462600250

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32780-64-6
Molecular Formula C19H25ClN2O3
Molecular Weight (g/mol) 364.87
InChI Key WQVZLXWQESQGIF-UHFFFAOYNA-N
IUPAC Name 2-hydroxy-5-{1-hydroxy-2-[(4-phenylbutan-2-yl)amino]ethyl}benzamide hydrochloride
SMILES Cl.CC(CCC1=CC=CC=C1)NCC(O)C1=CC=C(O)C(=C1)C(N)=O

Specifications

Color Colorless to Light Yellow-Pink
Assay Percent Range 98.5 % min. (HPLC)
Quantity 25 g
Formula Weight 364.87
Chemical Name or Material Laβlol hydrochloride
EINECSNumber 251-211-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.