Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labetalol Hydrochloride, USP, 97.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18603669 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-669 5 g Amber Glass Bottle
18-603-670 25 g Amber Glass Bottle
18-603-671 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-669 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L10695GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

L1069, 32780-64-6, C19H24N2O3·HCl

Spectrum™ Chemical Labetalol Hydrochloride, USP is a mixed alpha/beta adrenergic antagonist used in the treatment of high blood pressure. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 32780-64-6
Molecular Formula C19H25ClN2O3
Molecular Weight (g/mol) 364.87
InChI Key WQVZLXWQESQGIF-UHFFFAOYNA-N
IUPAC Name 2-hydroxy-5-{1-hydroxy-2-[(4-phenylbutan-2-yl)amino]ethyl}benzamide hydrochloride
SMILES Cl.CC(CCC1=CC=CC=C1)NCC(O)C1=CC=C(O)C(=C1)C(N)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 97.5 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.