Learn More
Cayman Chemical Labetalol (hydrochloride), Purity: ≥98%, CAS: 32780-64-6, 10 mg
SDP
Catalog No. 502935205
Encompass_Preferred
An analytical reference standard categorized as a β-adrenergic receptor antagonist; has been used as a doping agent in sports; intended for use in analytical forensic applications
Catalog No. 50-293-5205 Supplier Cayman Chemical Supplier No. 2628910 MG
An analytical reference standard categorized as a β-adrenergic receptor antagonist; has been used as a doping agent in sports; intended for use in analytical forensic applications