Learn More
Cayman Chemical Labetalol (hydrochloride), Purity: ≥98%, CAS: 32780-64-6, 50 mg
SDP
Catalog No. 502936993
Encompass_Preferred
An analytical reference standard categorized as a β-adrenergic receptor antagonist; has been used as a doping agent in sports; intended for use in analytical forensic applications
Catalog No. 50-293-6993 Supplier Cayman Chemical Supplier No. 2628950 MG
An analytical reference standard categorized as a β-adrenergic receptor antagonist; has been used as a doping agent in sports; intended for use in analytical forensic applications